Learn More
Zinc acetate dihydrate, 97+%
Description
Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in water and alcohol.
Notes
Incompatible with zinc salts, alkalies-their carbonates, oxalates, phosphates and sulfides.
Specifications
Specifications
| Melting Point | 237°C |
| Density | 1.735 |
| pH | 6 to 7 |
| Odor | Weak, Characteristic |
| Quantity | 100 g |
| UN Number | UN3077 |
| Beilstein | 3732513 |
| Merck Index | 14,10128 |
| Solubility Information | Soluble in water and alcohol. |
| SMILES | CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2] |
| Show More |
Safety and Handling
| Hazard Category | H302-H319 |
| Hazard Statement | GHS H Statement H302-H319 Harmful if swallowed. Causes serious eye irritation. |
| Precautionary Statement | P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 209-170-2 |
| RTECSNumber | ZG8750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only