missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate, 2 N (220 g + 870 mL Water), Ricca Chemical

Catalog No. R94500004A
Change view
Click to view available options
Quantity:
120 mL
500 mL
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R94500004A 4 L Natural Poly Bottle
94504 120 mL Natural Poly Bottle
945016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R94500004A Supplier Ricca Chemical Company Supplier No. R94500004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA for Sulfide Testing

Chemical Identifiers

Molecular Formula C4H10O6Zn
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Color Colorless
CAS 5970-45-6, 7732-18-5
Solubility Information Solubility in Water: Infinite
Molecular Weight (g/mol) 219.498
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼5 to 6
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Zinc Acetate, 2N (220g + 870mL Water)
Show More Show Less