missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-Mercaptobenzothiazole 98.0+%, TCI America™

$65.24 - $253.51

Chemical Identifiers

CAS 155-04-4
Molecular Formula C7H4NS2Zn+
Molecular Weight (g/mol) 231.616
MDL Number MFCD00072234
InChI Key JLKZUNOWPPEBPX-UHFFFAOYSA-M
Synonym 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate
PubChem CID 25200224
IUPAC Name zinc;1,3-benzothiazole-2-thiolate
SMILES C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
Z003325G
View Documents
TCI America
Z003325G
25 g N/A
Only null left
 
Z0033500G
View Documents
TCI America
Z0033500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

155-04-4
231.616
JLKZUNOWPPEBPX-UHFFFAOYSA-M
25200224
C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]
C7H4NS2Zn+
MFCD00072234
2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate
zinc;1,3-benzothiazole-2-thiolate

Specifications

155-04-4
White-Yellow
MFCD00072234
2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate
C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]
231.616
397.85
Crystalline Powder
325°C
C7H4NS2Zn+
25 g
JLKZUNOWPPEBPX-UHFFFAOYSA-M
zinc;1,3-benzothiazole-2-thiolate
25200224
≥98.0% (HPLC,T)
Zinc 2-Mercaptobenzothiazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-3418

RTECSNumber : DL7000000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'