CAS RN 155-04-4
IUPAC Name:
zinc(2+) bis((1,3-benzothiazol-2-yl)sulfanide)
Synonyms:
zinc(2+) bis(1,3-benzothiazol-2-ylsulfanide)
Molecular Weight (g/mol):
397.85
Molecular Formula:
C14H8N2S4Zn
InChi Key:
PGNWIWKMXVDXHP-UHFFFAOYSA-L
SMILES:
[Zn++].[S-]C1=NC2=CC=CC=C2S1.[S-]C1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
Zinc 2-Mercaptobenzothiazole 98.0+%, TCI America™
CAS: 155-04-4 Molecular Formula: C7H4NS2Zn+ Molecular Weight (g/mol): 231.616 MDL Number: MFCD00072234 InChI Key: JLKZUNOWPPEBPX-UHFFFAOYSA-M Synonym: 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate PubChem CID: 25200224 IUPAC Name: zinc;1,3-benzothiazole-2-thiolate SMILES: C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]