missing translation for 'onlineSavingsMsg'
Learn More

Zinc 2-Mercaptobenzothiazole 98.0+%, TCI America™

Catalog No. Z0033500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0033500G 500 g
Z003325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0033500G Supplier TCI America Supplier No. Z0033500G
Only null left

Chemical Identifiers

CAS 155-04-4
Molecular Formula C7H4NS2Zn+
Molecular Weight (g/mol) 231.616
MDL Number MFCD00072234
InChI Key JLKZUNOWPPEBPX-UHFFFAOYSA-M
Synonym 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate
PubChem CID 25200224
IUPAC Name zinc;1,3-benzothiazole-2-thiolate
SMILES C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]

Specifications

Melting Point 325°C
Color White-Yellow
Quantity 500 g
Formula Weight 397.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc 2-Mercaptobenzothiazole
EINECSNumber (5)-3418
RTECSNumber DL7000000
TSCA Yes