missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZIC3 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA5148993
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.Chemical Identifiers
| ZIC3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human, Mouse | |
| Zic3 | |
| Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203 | |
| Zic3 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| PBS with 5% trehalose and no preservative | |
| O60481, Q62521 | |
| 22773, 7547 | |
| E. coli-derived recombinant human ZIC3, His380-Val467. | |
| Antibody |
Specifications
| ZIC3 | |
| Polyclonal | |
| Unconjugated | |
| Zic3 | |
| Bn; heterotaxy 1; HTX; HTX1; Ka; RGD1561261; VACTERLX; ZIC; Zic family member 3; Zic family member 3 (odd-paired homolog, Drosophila); ZIC1; ZIC2; Zic3; Zinc finger protein 203; zinc finger protein of the cerebellum 3; Zinc finger protein ZIC 3; ZNF201; ZNF203 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 22773, 7547 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O60481, Q62521 | |
| Zic3 | |
| E. coli-derived recombinant human ZIC3, His380-Val467. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |