missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZHX3 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor.
Specifications
Specifications
| Antigen | ZHX3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | ZHX3 |
| Gene Accession No. | Q9H4I2 |
| Gene Alias | ZHX3; KIAA0395; TIX1 |
| Gene Symbols | ZHX3 |
| Host Species | Rabbit |
| Show More |