missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZHX2 Recombinant Rabbit Monoclonal Antibody (23GB4270)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553725
Description
ZHX2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
Zinc-fingers and homeobox (ZHX) proteins are transcription factors that interact with the activation domain of the A subunit of nuclear factor-Y (NF-YA). ZHX1-3 are ubiquitously expressed proteins expressed in various tissues. They act as transcriptional repressors and localize to the nucleus. The ZHX proteins contain two Cys(2)-His(2)-type zinc-finger motifs and five homeodomains (HDs). These domains allow the ZHX proteins to form homodimers, but they can also form heterodimers with each other. However, this dimerization is not required for repressor activity. Hypermethylation-mediated silencing of ZHX2 is an epigenetic event involved in hepatocellular carcinoma (HCC).Chemical Identifiers
| ZHX2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ZHX2 | |
| AFP regulator 1; AFR1; Afr-1; alpha fetoprotein regulation 1, adult; alpha-fetoprotein regulator 1; KIAA0854; LOW QUALITY PROTEIN: zinc fingers and homeoboxes protein 2; mKIAA0854; Raf; Regulator of AFP; transcription factor ZHX2; Zhx2; Zinc finger and homeodomain protein 2; zinc fingers and homeoboxes 2; zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2 | |
| ZHX2 | |
| Primary | |
| Affinity chromatography |
| 23GB4270 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80VX4, Q8C0C0, Q9Y6X8 | |
| 22882, 314988, 387609 | |
| A synthesized peptide derived from human ZHX2 (260-500AA). | |
| Antibody |
Specifications
| ZHX2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ZHX2 | |
| AFP regulator 1; AFR1; Afr-1; alpha fetoprotein regulation 1, adult; alpha-fetoprotein regulator 1; KIAA0854; LOW QUALITY PROTEIN: zinc fingers and homeoboxes protein 2; mKIAA0854; Raf; Regulator of AFP; transcription factor ZHX2; Zhx2; Zinc finger and homeodomain protein 2; zinc fingers and homeoboxes 2; zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22882, 314988, 387609 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4270 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80VX4, Q8C0C0, Q9Y6X8 | |
| ZHX2 | |
| A synthesized peptide derived from human ZHX2 (260-500AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |