missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenon™ pHrodo™ iFL IgG Labeling Reagents
GREENER_CHOICE

Catalog No. Z25626
Change view
Click to view available options
Quantity:
250 μL
1.2 mL
Species:
Human
Mouse
Label or Dye:
pHrodo Deep Red
pHrodo Green
pHrodo iFL Green
pHrodo iFL Red
pHrodo Red
Color:
Deep Red
Green
Red
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Species Label or Dye Color
Z25626 1.2 mL Mouse pHrodo Red Red
Z25618 250 μL Human pHrodo Deep Red Deep Red
Z25622 250 μL Mouse pHrodo Deep Red Deep Red
Z25612 250 μL Human pHrodo iFL Red Red
Z25611 250 μL Human pHrodo iFL Green Green
Z25610 250 μL Mouse pHrodo iFL Red Red
Z25609 250 μL Mouse pHrodo iFL Green Green
Z25623 1.2 mL Human pHrodo Deep Red Deep Red
Z25614 1.2 mL Human pHrodo iFL Red Red
Z25613 1.2 mL Human pHrodo iFL Green Green
Z25624 1.2 mL Mouse pHrodo Deep Red Deep Red
Z25625 1.2 mL Mouse pHrodo Green Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. Z25626 Supplier Invitrogen™ Supplier No. Z25626

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex.

Streamline antibody-drug conjugate screening campaigns with Invitrogen Zenon pHrodo IgG Labeling Reagents to screen for antibody internalization. These reagents provide rapid, scalable antibody labeling and give a bright fluorescence-based indication of antibody uptake and trafficking in target cells when analyzed by flow cytometry, fluorescence microscopy, or high-content screening.

Zenon™ pHrodo™ iFL Green-labeled or Red-labeled Fab fragments (i.e. labeling reagent) are directed against the Fc portion of an intact IgG primary antibody to form a labeling complex. Formation of the Fab–antibody complex occurs in fewer than 5 minutes. Because the pHrodo™ iFL dyes dramatically increase in fluorescence as the pH of their surroundings becomes more acidic, the antibodies coupled with the Zenon™ pHrodo™ iFL IgG Labeling Reagents provide an excellent measure of endocytic activity based on the acidification of the labeled antibodies as they are ingested by the cell.

The Zenon™ pHrodo™ iFL IgG Labeling Reagents contain sufficient material to label one to four 96-well plates, depending on the Zenon™ labeling reagent concentration used. Only a small amount of starting material is needed, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions.

Other advantages of the pH-sensitive Zenon™ pHrodo™ iFL IgG Labeling Reagents include:

  • Simplicity—No reagent purification is required
  • Accuracy—No wash steps or quenchers are needed, minimizing cross-reactivity or background
  • Speed—Labeled antibodies are typically ready to use in 10 minutes
  • Specificity—Dyes are designed for the specific detection of phagocytosis and endocytosis

Preserve primary antibody function and affinities

Reactive dye labeling of primary antibodies can have unpredictable and undesirable outcomes. One of these is reduced binding affinity due to label addition in the binding pocket. The Zenon™ antibody labeling approach, which is targeted to the Fc tail, resolves this potential issue.

Try custom antibody conjugates

If the antibody conjugate you need is not available as one of our stocked products, we will prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Store in refrigerator at 2°C to 8°C.

Specifications

For Use With (Application) To screen for antibody internalization
Product Type Labeling Reagent
Quantity 1.2 mL
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Detection Method Fluorescence
Excitation/Emission 560/585 nm
For Use With (Equipment) Flow Cytometer, High Content Analysis, IncuCyte, Fluorescence Microscope
Labeling Method Rapid, no wash
Product Line Labeling Kits
Solubility Aqueous Buffer
Label or Dye pHrodo Red
Color Red
Labeling Target Antibodies
Species Mouse
Label Type Mouse IgG Fc domain
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.