missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zeniquin™ Marbofloxacin, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| Product Line | Zeniquin |
| CAS | 115550-35-1 |
| Physical Form | Neat |
| Linear Formula | C17H19 FN4O4 |
| MDL Number | MFCD00864820 |
| Synonym | 9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-piperazino)-7-oxo-7 H-pyrido[1,2,3-ij][1,2,4 ]benzoxadiazine-6-carboxylic acid; Zeniquin™ |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 362.36 |
| Grade | Analytical Standard |
Zeniquin is a registered trademark of Pfizer, Inc.