missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebularine 98.0+%, TCI America™
$341.13 - $1010.32
Chemical Identifiers
| CAS | 3690-10-6 |
|---|---|
| Molecular Formula | C9H12N2O5 |
| Molecular Weight (g/mol) | 228.20 |
| MDL Number | MFCD04973699 |
| InChI Key | RPQZTTQVRYEKCR-UHFFFAOYNA-N |
| Synonym | zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl |
| PubChem CID | 100016 |
| ChEBI | CHEBI:46938 |
| IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| SMILES | C1=CN(C(=O)N=C1)C2C(C(C(O2)CO)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
Z0022200MG
|
TCI America
Z0022200MG |
200 mg |
Each for $341.13
|
|
|||||
|
Z00221G
|
TCI America
Z00221G |
1 g |
Each for $1,010.32
|
|
|||||
Chemical Identifiers
| 3690-10-6 | |
| 228.20 | |
| RPQZTTQVRYEKCR-UHFFFAOYNA-N | |
| 100016 | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| C9H12N2O5 | |
| MFCD04973699 | |
| zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl | |
| CHEBI:46938 | |
| C1=CN(C(=O)N=C1)C2C(C(C(O2)CO)O)O |
Specifications
| 3690-10-6 | |
| White-Yellow | |
| MFCD04973699 | |
| zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl | |
| C1=CN(C(=O)N=C1)C2C(C(C(O2)CO)O)O | |
| 228.20 | |
| CHEBI:46938 | |
| ≥98.0% (HPLC,T) | |
| Zebularine |
| 158°C | |
| C9H12N2O5 | |
| 200 mg | |
| RPQZTTQVRYEKCR-UHFFFAOYNA-N | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one | |
| 100016 | |
| 228.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UW7539720
TSCA : No
Recommended Storage : Refrigerator