missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL

Catalog No. FEA40002475
Change view
Click to view available options
Quantity:
1 column x 5 mL
10 Columns
2 Plates
25 Columns
4 Plates
5 Columns
5 columns x 1 mL
50 Columns
Format:
96-well Filter Plate
Chromatography Cartridge
Spin Column
Volume (Metric):
96-well
75 μL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Volume (Metric)
FEA40002475 10 Columns Spin Column 20 mL
PI89877 25 Columns Spin Column 75 μL
PI89878 50 Columns Spin Column 75 μL
FEA40002469 5 Columns Spin Column 20 mL
PI89890 25 Columns Spin Column 2 mL
PI89891 5 Columns Spin Column 5 mL
PI89892 25 Columns Spin Column 5 mL
PI89893 5 Columns Spin Column 10 mL
PI89894 25 Columns Spin Column 10 mL
PI89889 5 Columns Spin Column 2 mL
PI89882 25 Columns Spin Column 0.5 mL
PI89883 50 Columns Spin Column 0.5 mL
PI89934 5 columns x 1 mL Chromatography Cartridge 1 mL
PI89935 1 column x 5 mL Chromatography Cartridge 5 mL
PI89807 2 Plates 96-well Filter Plate 96-well
PI89808 4 Plates 96-well Filter Plate 96-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. FEA40002475 Supplier Thermo Scientific™ Supplier No. A40002475

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform high-throughput protein recovery (7K MWCO) and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Thermo Scientific Pierce Zeba desalting chromatography columns, plates, and cartridges.

Perform high-throughput protein recovery and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Pierce Zeba desalting chromatography columns, plates, and cartridges. Rapidly desalt protein and biological samples for manual or automated liquid chromatography (LC) with ≥95% retention of salts and other small molecular contaminants (<1,000 Da).

Zeba Spin Desalting columns, plates, and chromatography cartridge fittings are convenient, ready-to-use devices compatible with major automated liquid-chromatography systems or for manual syringe processing. The polypropylene columns have a 7K MWCO and contain a proprietary high-performance size-exclusion chromatography resin (Zeba High-Performance Resin) that offers excellent protein desalting and recovery in a centrifuge format. When using Zeba Spin Desalting Columns, even very dilute (25 μg/mL) protein samples can be successfully processed to obtain greater than 95% retention (removal) of salts and other small molecules (<1000 MW), and good recovery of proteins and other macromolecules (>7000 MW).

Similarly, Zeba Desalting plates (7K MWCO) are polypropylene devices that contain Zeba High-Performance Resin and provide excellent protein desalting performance, with high recovery of proteins >7000 Da in a centrifuge format for small sample volumes. The plates enable fast, consistent, well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations. Zeba 96-well spin desalting plates require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in just five minutes, offering rapid and exceptional protein desalting.

The Zeba Desalting Chromatography Cartridges contain Zeba High-Performance Resin for desalting or buffer exchange applications. This resin is ideal for removing low molecular weight compounds including salts, fluorescent dyes, biotin, and other small labeling reagents. Protein samples can be processed with unsurpassed protein recovery and 95% retention of salts and other small molecular contaminants (<1,000 Da). Zeba Desalting cartridges provide fast, easy, and reproducible chromatographic separations and can be regenerated for multiple uses.

Benefits of Zeba Spin Desalting columns, plates, and cartridges include:

  • High recovery—low-binding resin maximizes protein recovery
  • Fast—no fraction screening or waiting for protein to emerge by gravity flow
  • Economical—great performance at lower cost than other commercially available cartridges
  • Easy-to-use—no cumbersome column preparation or equilibration
  • Flexible—available in spin columns, filter spin plates and cartridges for a range of needs

Available formats

  • Spin columns—75 μL, 0.5 mL (microcentrifuge), 2 mL, 5 mL, 10 mL columns
  • 96-well spin plates—pre-dispensed 96-well filter plates, compatible with centrifugation for manual or automated purification
  • Chromatography cartridges—compatible with automated liquid chromatography systems and with syringe processing

Specifications

Description Zeba™ Spin Desalting Columns, 7K MWCO, 10 Columns
Formulation 20 mL Zeba Desalting Resin Bed
Product Type Spin Desalting Columns
For Use With (Equipment) Standard Table-top Centrifuge
For Use With (Application) Small molecule removal and buffer exchange
Content And Storage Store at 4°C
Format Spin Column
Purification Target Buffer Exchange, Protein
Column Type Size-exclusion, Proprietary Resin
MWCO 7.0 kDa
Product Line Zeba
Disposable Single-use
Quantity 10 Columns
Volume (Metric) 20 mL
Sample Volume (Metric) 4 to 10 mL
Show More Show Less
Can Zeba Desalting columns, plates, and cartridges be autoclaved?

Yes, Zeba Desalting columns, plates, and cartridges can be autoclaved at 115 degrees C for 30 min.

What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?

Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)

How should I store Zeba Spin Desalting Columns, Plates, and Cartridges?

We recommend storing Zeba Spin Desalting Columns, Plates, and Cartridges at 4 degrees C.