missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zeaxanthin, ≥95% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 144-68-3 |
| Physical Form | Neat |
| Linear Formula | C40H56O2 |
| MDL Number | MFCD08435940 |
| Synonym | beta,beta-Carotene-3,3′-diol |
| Type | Analytical Standard |
| Quantity | 1 mg |
| Formula Weight | 568.87 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |