missing translation for 'onlineSavingsMsg'
Learn More

α-Zearalenol Solution, 10 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111017988
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017988 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017988 Supplier Merck Emd Millipore Supplier No. 354061ML
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 36455-72-8
Flash Point 2.0°C - Closed Cup
Linear Formula C18H24O5
MDL Number MFCD16657008
Synonym 2,4-Dihydroxy-6-(6 alpha,10-dihydroxy-trans-1-undecenyl)benzoic acid mu-lactone solution
Type Analytical Standard
Quantity 1 mL
Formula Weight 320.38
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetonitrile