missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553957
Description
Immunogen sequence: SACTVKTGLD PTLVGICGEG TESVQSLLLG AVPKGNATKE YMESLQLKPG EVIYKCPKCC Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 52%.
ZDHHC7 is a protein coding gene. Palmitoyltransferase with broad specificity. Palmitoylates JAM3. Palmitoylates SNAP25 and DLG4/PSD95. Palmitoylates sex steroid hormone receptors, including ESR1, PGR and AR, thereby regulating their targeting to the plasma membrane and their function in rapid intracellular signaling upon binding of sex hormones. May play a role in follicle stimulation hormone (FSH) activation of testicular Sertoli cells.Chemical Identifiers
| ZDHHC7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ZDHHC7 | |
| AL024087; DHHC7; DHHC-7; GABA-A receptor-associated membrane protein 2; Gramp2; membrane-associated DHHC7 zinc finger protein; palmitoyltransferase ZDHHC7; putative zinc finger protein SERZ-1; sertoli cell gene with a zinc finger domain protein; Sertoli cell gene with zinc finger domain-β; Serz; SERZ1; Serz-1; SERZ-B; Zdhhc7; Zinc finger DHHC domain-containing protein 7; zinc finger DHHC-type containing 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370 | |
| ZDHHC7 | |
| Primary | |
| Antigen affinity chromatography |
| 0.05 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NXF8 | |
| 55625 | |
| Recombinant protein corresponding to Human ZDHHC7. Recombinant protein control fragment (Product #RP-91160). | |
| Antibody |
Specifications
| ZDHHC7 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC7 | |
| AL024087; DHHC7; DHHC-7; GABA-A receptor-associated membrane protein 2; Gramp2; membrane-associated DHHC7 zinc finger protein; palmitoyltransferase ZDHHC7; putative zinc finger protein SERZ-1; sertoli cell gene with a zinc finger domain protein; Sertoli cell gene with zinc finger domain-β; Serz; SERZ1; Serz-1; SERZ-B; Zdhhc7; Zinc finger DHHC domain-containing protein 7; zinc finger DHHC-type containing 7; zinc finger protein 370; zinc finger, DHHC domain containing 7; zinc finger, DHHC-type containing 7; ZNF370 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NXF8 | |
| ZDHHC7 | |
| Recombinant protein corresponding to Human ZDHHC7. Recombinant protein control fragment (Product #RP-91160). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |