missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113122
Description
ZDHHC23 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Palmitoyltransferase that mediates palmitoylation of KCNMA1, regulating localization of KCNMA1 to the plasma membrane. May be involved in NOS1 regulation and targeting to the synaptic membrane.Chemical Identifiers
| ZDHHC23 | |
| -20°C or -80°C if preferred | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| ZDHHC23 | |
| DHHC-23; DHHC-containing protein 11; Gm1751; Gm779; NIDD; nNOS-intereacting DHHC-containing Dem protein-L; palmitoyltransferase ZDHHC23; probable palmitoyltransferase ZDHHC23; Zdhhc23; Zinc finger DHHC domain-containing protein 23; zinc finger DHHC-type containing 23; zinc finger, DHHC domain containing 23 | |
| ZDHHC23 | |
| Primary | |
| Protein G |
| 1 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5Y5T3, Q8IYP9 | |
| 254887, 332175 | |
| Recombinant Human Palmitoyltransferase ZDHHC23 protein (187-302AA). | |
| Antibody |
Specifications
| ZDHHC23 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC23 | |
| DHHC-23; DHHC-containing protein 11; Gm1751; Gm779; NIDD; nNOS-intereacting DHHC-containing Dem protein-L; palmitoyltransferase ZDHHC23; probable palmitoyltransferase ZDHHC23; Zdhhc23; Zinc finger DHHC domain-containing protein 23; zinc finger DHHC-type containing 23; zinc finger, DHHC domain containing 23 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 254887, 332175 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5Y5T3, Q8IYP9 | |
| ZDHHC23 | |
| Recombinant Human Palmitoyltransferase ZDHHC23 protein (187-302AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |