missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCWCC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
Specifications
Specifications
| Antigen | ZCWCC1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | MORC2 |
| Gene Accession No. | Q9Y6X9 |
| Gene Alias | MORC family CW-type zinc finger protein 2, Zinc finger CW-type coiled-coil domain protein 1, MORC2, KIAA0852, ZCWCC1 |
| Gene Symbols | MORC2 |
| Show More |