missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB17 Monoclonal Antibody (K49033_2B4), Invitrogen™
Mouse Monoclonal Antibody
$1461.54 - $2194.06
Specifications
| Antigen | ZBTB17 |
|---|---|
| Clone | K49033_2B4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: GSEVSVVTVD DMVTLATEAL AATAVTQLTV VPVGAAVTAD ETEVLKAEIS KAVKQVQEED PNTHILYACD SCGDKFLDAN SLAQHVRIHT AQALVMFQTD
ZBTB17 Monoclonal antibody specifically detects ZBTB17 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZBTB17 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60 | |
| Recombinant human ZBTB17 (immunogen range is 651-750). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| ZBTB17 |
| K49033_2B4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q13105, Q60821 | |
| 22642, 313666, 7709 | |
| IgG1 | |
| Protein A | |
| Antibody |