missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZBTB17 Monoclonal Antibody (K49033_2B4), Invitrogen™

Catalog No. MA556002
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556002 200 μL
MA555622 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556002 Supplier Invitrogen™ Supplier No. MA556002
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GSEVSVVTVD DMVTLATEAL AATAVTQLTV VPVGAAVTAD ETEVLKAEIS KAVKQVQEED PNTHILYACD SCGDKFLDAN SLAQHVRIHT AQALVMFQTD

Transcription factor that can function as an activator or repressor depending on its binding partners, and by targeting negative regulators of cell cycle progression. Plays a critical role in early lymphocyte development, where it is essential to prevent apoptosis in lymphoid precursors, allowing them to survive in response to IL7 and undergo proper lineage commitment. Has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. Required for early embryonic development during gastrulation.

Specifications

Antigen ZBTB17
Applications Western Blot
Classification Monoclonal
Clone K49033_2B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene ZBTB17
Gene Accession No. Q13105, Q60821
Gene Alias AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60
Gene Symbols ZBTB17
Host Species Mouse
Immunogen Recombinant human ZBTB17 (immunogen range is 651-750).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22642, 313666, 7709
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less