missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB17 Monoclonal Antibody (K49033_2B4), Invitrogen™
Description
Sequence of this protein is as follows: GSEVSVVTVD DMVTLATEAL AATAVTQLTV VPVGAAVTAD ETEVLKAEIS KAVKQVQEED PNTHILYACD SCGDKFLDAN SLAQHVRIHT AQALVMFQTD
Transcription factor that can function as an activator or repressor depending on its binding partners, and by targeting negative regulators of cell cycle progression. Plays a critical role in early lymphocyte development, where it is essential to prevent apoptosis in lymphoid precursors, allowing them to survive in response to IL7 and undergo proper lineage commitment. Has been shown to bind to the promoters of adenovirus major late protein and cyclin D1 and activate transcription. Required for early embryonic development during gastrulation.
Specifications
Specifications
| Antigen | ZBTB17 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | K49033_2B4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide |
| Gene | ZBTB17 |
| Gene Accession No. | Q13105, Q60821 |
| Gene Alias | AA589413; LP-1; Miz1; Miz-1; Myc-interacting zinc finger protein 1; Myc-interacting Zn finger protein-1; mZ13; pHZ-67; Polyomavirus late initiator promoter-binding protein; ZBTB17; Zfp100; Zfp-100; zinc finger and BTB domain containing 17; zinc finger and BTB domain-containing protein 17; Zinc finger protein 100; zinc finger protein 151; zinc finger protein 151 (pHZ-67); zinc finger protein 60; Zinc finger protein Z13; ZNF151; ZNF60 |
| Show More |