missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBP1, Mouse anti-Human, Clone: Zippy-1, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB16085
Specifications
| ZBP1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| C20orf183/DAI/DLM-1/DLM1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| Zippy-1 | |
| Immunoprecipitation (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Z-DNA binding protein 1 | |
| ZBP1 | |
| Recombinant protein corresponding to amino acids 1-411 of mouse ZBP1. | |
| RUO | |
| 81030 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |