missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZAK, GST Tag Recombinant Protein
Recombinant Protein
$872.00 - $21850.00
Specifications
| Accession Number | Q9NYL2 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with no preservative; pH 7.5 |
| Gene ID (Entrez) | 51776 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7052A
|
Invitrogen™
PR7052A |
100 μg |
Each for $3,095.00
|
|
|||||
PV4200
|
Invitrogen™
PV4200 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PV3882
|
Invitrogen™
PV3882 |
10 μg |
Each for $872.00
|
|
|||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
ZAK is a leucine zipper-containing mitogen-activated protein related to the MLKs in both sequence and function. Like the MLKs and other MAP3Ks, ZAK activates MAP2Ks. ZAK is a member of the MAPKKK family of signal transduction molecules and encodes a protein with an N-terminal kinase catalytic domain, followed by a leucine zipper motif and a sterile-alpha motif. This magnesium-binding protein forms homodimers and is located in the cytoplasm. The protein mediates gamma radiation signaling leading to cell cycle arrest and activity of this protein plays a role in cell cycle checkpoint regulation in cells. The protein also has pro-apoptotic activity. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| Q9NYL2 | |
| Kinase Assay | |
| 51776 | |
| Human ZAK, GST Tag | |
| ZAK | |
| Human | |
| GST-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| See Label | |
| 50 mM tris with no preservative; pH 7.5 | |
| 80.8 kDa | |
| AV006891; AZK; B230120H23Rik; HCCS4; HCCS-4; Human cervical cancer suppressor gene 4 protein; leucine zipper- and sterile alpha motif kinase ZAK; Leucine zipper- and sterile alpha motif-containing kinase; Map3k20; Mitogen-activated protein kinase kinase kinase 20; Mitogen-activated protein kinase kinase kinase MLT; mixed lineage kinase 7; mixed lineage kinase with a leucine zipper and a sterile alpha motif; mixed lineage kinase-related kinase; mixed lineage kinase-related kinase MRK-beta; MLK7; mlklak; MLK-like mitogen-activated protein triple kinase; MLK-related kinase; MLT; MLTK; MLTKalpha; MLTKbeta; MRK; Pk.; RGD1561394; SFMMP; sterile alpha motif and leucine zipper containing kinase AZK; sterile alpha motif- and leucine zipper-containing kinase AZK; sterile-alpha motif and leucine zipper containing kinase AZK; ZAK | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Insect cells |