missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Z-Val-Val-Nle Diazomethyl Ketone

Catalog No. aaj64544mf
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64544MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64544MF Supplier Thermo Scientific Chemicals Supplier No. J64544MF

CAS: 155026-49-6 | C25H37N5O5, C25H37N5O5 | 487.60 g/mol

Z-Val-Val-Nle diazomethyl ketone is used as an irreversible cathepsin inhibitor that is 380 times more effective in inactivating cathepsin S than L. It is often abbreviated as DMK. Peptidyl diazomethyl ketones are specific inactivators of thiol proteinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 155026-49-6
Molecular Formula C25H37N5O5
Molecular Weight (g/mol) 487.60
InChI Key IIPZPLSBQVJCBO-UHFFFAOYNA-N
Synonym C-3890; Z-Val-Val-Nle-DMK
IUPAC Name benzyl N-[1-({1-[(1-diazo-2-oxoheptan-3-yl)carbamoyl]-2-methylpropyl}carbamoyl)-2-methylpropyl]carbamate
SMILES CCCCC(NC(=O)C(NC(=O)C(NC(=O)OCC1=CC=CC=C1)C(C)C)C(C)C)C(=O)C=[N+]=[N-]

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inactivates thiol proteinases; an irreversible cathepsin inhibitor
Formula Weight 487.60g/mol
Product Type Signal Transduction Reagent
Form Powder
Recommended Storage Store at -20°C

RUO – Research Use Only