missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 6 Peptide
Z'-LYTE Kinase Assay Kit - Ser/Thr 6 Peptide
Supplier: Invitrogen™ PV3179
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide contains an R-X-R-X-X-S-Hyd consensus sequence, where Hyd is a bulky hydrophobic residue, and X is any residue.Specifications
| Z´-LYTE | |
| FRET | |
| Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
| Methylcoumarin | |
| 800 x 20 μL Assays | |
| 405⁄465, 535 | |
| End Point | |
| Signal Transduction | |
| SGK, SGK2, RPS6KA3, AKT2, RPS6KA1, AKT1, CLK2, PDK1, RPS6KA2, SGKL, AKT3 |
| Serine/Threonine Kinases | |
| Serine/Threonine Kinase Assay | |
| Biochemical phospho-specific activity (no antibody) | |
| Kinase Assay Kit | |
| 384-well plate | |
| 800 Reactions | |
| Dry Ice | |
| Peptide-Based Substrate | |
| AKT1, AKT2, AKT3, CLK2, PDK1, RPS6KA1, RPS6KA2, RPS6KA3, SGK, SGK2, SGKL |
For Research Use Only. Not for use in diagnostic procedures.