missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 1 Peptide
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon a porcine pyruvate liver kinase derived peptide used to study PKA substrate specificity, and containing an R-R-X-S consensus sequence, where X is any residue.
Specifications
Specifications
| Target | Serine/Threonine Kinases |
| For Use With (Application) | FRET |
| Assay | Serine/Threonine Kinase Assay |
| Content And Storage | Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Label or Dye | Methylcoumarin |
| Product Type | Kinase Assay Kit |
| Quantity | 800 x 20 μL Assays |
| Format | 384-well plate |
| Sample Type | porcine pyruvate liver kinase (LRRASLG) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.