missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-2-Chloro-2-butene 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | -106°C |
| Color | Yellow |
| Boiling Point | 72°C |
| Quantity | 5 mL |
| UN Number | 1993 |
| Formula Weight | 90.55 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (Z)-2-Chloro-2-butene |
Safety and Handling
| EINECSNumber | (2)-0117 |
| RTECSNumber | EM4265000 |
| TSCA | No |