accessibility menu, dialog, popup

UserName

CH3H3CCl(E)

CAS RN 2211-69-0

IUPAC Name: (2E)-2-chlorobut-2-ene
Synonyms: 2-butene, 2-chloro-
Molecular Weight (g/mol): 90.55
Molecular Formula: C4H7Cl
InChi Key: DSDHFHLZEFQSFM-ONEGZZNKSA-N
SMILES: C\C=C(/C)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results