missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7137024
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19215G 5 g
T192125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19215G Supplier TCI America Supplier No. T19215G

Chemical Identifiers

CAS 52093-30-8
Molecular Formula C3F9O9S3Y
Molecular Weight (g/mol) 536.10
MDL Number MFCD00209623
InChI Key JPJIEXKLJOWQQK-UHFFFAOYSA-K
Synonym yttrium iii trifluoromethanesulfonate, yttrium triflate, yttrium iii triflate, yttrium 3+ tritriflate, trifluoromethanesulfonate; yttrium 3+, yttrium trifluoromethanesulfonate, trifluoromethanesulfonic acid yttrium iii salt, acmc-209kxu, yttrium trifluoromethanesulfonate?
PubChem CID 2733939
IUPAC Name yttrium(3+) tritrifluoromethanesulfonate
SMILES [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 536.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Yttrium(III) Trifluoromethanesulfonate