Learn More
Yttrium(III) trifluoromethanesulfonate hydrate
CAS: 34629-25-9 | C3F9O9S3Y | 536.10 g/mol
$58.97 - $165.22
Chemical Identifiers
| CAS | 34629-25-9 |
|---|---|
| Molecular Formula | C3F9O9S3Y |
| Molecular Weight (g/mol) | 536.10 |
| MDL Number | MFCD00209623 |
| InChI Key | JPJIEXKLJOWQQK-UHFFFAOYSA-K |
| Synonym | yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:? |
| PubChem CID | 57349102 |
| SMILES | [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
Description
It is employed in preparation of superconducting thin films. It is used in chemical research & as a intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 34629-25-9 | |
| 536.10 | |
| JPJIEXKLJOWQQK-UHFFFAOYSA-K | |
| 57349102 |
| C3F9O9S3Y | |
| MFCD00209623 | |
| yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:? | |
| [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
Specifications
| 34629-25-9 | |
| 2 g | |
| MFCD00209623 | |
| JPJIEXKLJOWQQK-UHFFFAOYSA-K | |
| yttrium(3+) tritrifluoromethanesulfonate | |
| 57349102 | |
| Yttrium(III) trifluoromethanesulfonate hydrate |
| Powder | |
| C3F9O9S3Y | |
| yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:? | |
| [Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F | |
| 536.10 | |
| 536.10 (Anhydrous) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only