missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) trifluoromethanesulfonate hydrate

CAS: 34629-25-9 | C3F9O9S3Y | 536.10 g/mol

Supplier:  Thermo Scientific Chemicals 04031509

 View more versions of this product

Catalog No. AA4031509

Only null left
Explore available promotions

Description

Description

It is employed in preparation of superconducting thin films. It is used in chemical research & as a intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Yttrium(III) trifluoromethanesulfonate hydrate
Powder
C3F9O9S3Y
yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:?
[Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
536.10
536.10 (Anhydrous)
34629-25-9
10 g
MFCD00209623
JPJIEXKLJOWQQK-UHFFFAOYSA-K
yttrium(3+) tritrifluoromethanesulfonate
57349102

Chemical Identifiers

34629-25-9
536.10
JPJIEXKLJOWQQK-UHFFFAOYSA-K
57349102
C3F9O9S3Y
MFCD00209623
yttrium iii trifluoromethanesulfonate hydrate, acmc-20aj8x, 3cf3o3s.y.h2o, yttrium 3+ hydrate tritriflate, yttrium trifluoromethanesulfonate-water 1/3/1, methanesulfonic acid,1,1,1-trifluoro-, yttrium 3+ salt, hydrate 3:1:?
[Y+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only