missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Color | White |
| Physical Form | Adhering Crystalline Powder or Crystals |
| Quantity | 500 g |
| Assay Percent Range | 99.9% |
| Linear Formula | Y(NO3)3·6H2O |
| Merck Index | 14,10107 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol |
| Formula Weight | 383.01 |
| Percent Purity | 99.9% |
| Grade | Trace Metal Basis |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye irritation. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medic |
| RTECSNumber | ZG3750000 |
RUO – Research Use Only