missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) Isopropoxide 98.0+%, TCI America™

Catalog No. p-7137984
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y0009200MG 200 mg
Y00091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y0009200MG Supplier TCI America Supplier No. Y0009200MG
Only null left

Chemical Identifiers

CAS 2172-12-5
Molecular Formula C9H24O3Y
Molecular Weight (g/mol) 269.19
MDL Number MFCD00015641
InChI Key NREVZTYRXVBFAQ-UHFFFAOYSA-N
Synonym yttrium isopropoxide
PubChem CID 85308566
IUPAC Name propan-2-ol;yttrium
SMILES CC(C)O.CC(C)O.CC(C)O.[Y]

Specifications

Melting Point 290°C
Color White
Quantity 200 mg
UN Number 1325
Formula Weight 266.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Yttrium(III) Isopropoxide