missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium AA Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical
$162.22 - $162.22
Chemical Identifiers
| CAS | 1314-36-9 |
|---|---|
| Molecular Formula | O3Y2 |
| Molecular Weight (g/mol) | 225.81 |
| MDL Number | MFCD00011473 |
| InChI Key | RUDFQVOCFDJEEF-UHFFFAOYSA-N |
| IUPAC Name | diyttrium(3+) trioxidandiide |
| SMILES | [O--].[O--].[O--].[Y+3].[Y+3] |
Chemical Identifiers
| 1314-36-9 | |
| 225.81 | |
| RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
| [O--].[O--].[O--].[Y+3].[Y+3] |
| O3Y2 | |
| MFCD00011473 | |
| diyttrium(3+) trioxidandiide |
Specifications
| Natural LDPE Bottle | |
| 93.0,4.8,0.12 | |
| Liquid | |
| MFCD00011473 | |
| < 2 | |
| Standard | |
| [O--].[O--].[O--].[Y+3].[Y+3] | |
| 225.81 | |
| 1mL = 1mg Y (1, 000ppm Y)Y2O3 in 3% HNO3 |
| 1314-36-9,7697-37-2,7732-18-5 | |
| 97.0,4.99,0.12 | |
| O3Y2 | |
| Colorless | |
| Approximately 0°C | |
| RUDFQVOCFDJEEF-UHFFFAOYSA-N | |
| diyttrium(3+) trioxidandiide | |
| 100 mL | |
| Yttrium AA Standard |
Safety and Handling
ShelfLife : 24 months