missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium(III) Trifluoromethanesulfonate Hydrate 98.0+%, TCI America™
Supplier: TCI America T161025G
Specifications
| Ytterbium(III) Trifluoromethanesulfonate Hydrate | |
| White-Yellow | |
| MFCD03703499 | |
| 1759 | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| trifluoromethanesulfonate;ytterbium(3+) | |
| 2733225 | |
| ≥98.0% (T) |
| 54761-04-5 | |
| C3F9O9S3Yb | |
| 25 g | |
| ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3] | |
| 620.244 | |
| 620.24 | |
| Crystalline Powder |
Chemical Identifiers
| 54761-04-5 | |
| 620.244 | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| 2733225 | |
| C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3] |
| C3F9O9S3Yb | |
| MFCD03703499 | |
| ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate | |
| trifluoromethanesulfonate;ytterbium(3+) |
Safety and Handling
TSCA : No