missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) Trifluoromethanesulfonate Hydrate 98.0+%, TCI America™

Catalog No. p-7136872
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16105G 5 g
T161025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16105G Supplier TCI America Supplier No. T16105G
Only null left

Chemical Identifiers

CAS 54761-04-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.244
MDL Number MFCD03703499
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate, ytterbium iii triflate, ytterbium triflate, unii-4n70osw24g, ytterbium trifluoromethanesulfonate, yb otf 3, ytterbium 3+ tritriflate, trifluoromethanesulfonate; ytterbium 3+, ytterbium 3+ triflate, ytterbium trifluoromethanesulphonate
PubChem CID 2733225
IUPAC Name trifluoromethanesulfonate;ytterbium(3+)
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Yb+3]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 620.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ytterbium(III) Trifluoromethanesulfonate Hydrate