Learn More
Ytterbium(III) sulfate octahydrate, REacton™, 99.9% (REO)
CAS: 10034-98-7 | H16O20S3Yb2 | 778.40 g/mol
$100.99 - $464.27
Chemical Identifiers
| CAS | 10034-98-7 |
|---|---|
| Molecular Formula | H16O20S3Yb2 |
| Molecular Weight (g/mol) | 778.40 |
| MDL Number | MFCD00149887 |
| InChI Key | MDOSIOHMOWGVIM-UHFFFAOYSA-H |
| Synonym | unii-e5w6do92vs, e5w6do92vs, 3so4.2yb.8h2o, ytterbium sulfate,, ytterbium sulfate octahydrate, ytterbium sulfate, ytterbium iii sulfate octahydrate, reacton, diytterbium 3+ octahydrate trisulfate, sulfuric acid,ytterbium 3+ salt 3:2 , octahydrate 8ci,9ci |
| PubChem CID | 25022290 |
| SMILES | O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10034-98-7 | |
| 778.40 | |
| MDOSIOHMOWGVIM-UHFFFAOYSA-H | |
| 25022290 |
| H16O20S3Yb2 | |
| MFCD00149887 | |
| unii-e5w6do92vs, e5w6do92vs, 3so4.2yb.8h2o, ytterbium sulfate,, ytterbium sulfate octahydrate, ytterbium sulfate, ytterbium iii sulfate octahydrate, reacton, diytterbium 3+ octahydrate trisulfate, sulfuric acid,ytterbium 3+ salt 3:2 , octahydrate 8ci,9ci | |
| O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Specifications
| 10034-98-7 | |
| 10 g | |
| MFCD00149887 | |
| unii-e5w6do92vs, e5w6do92vs, 3so4.2yb.8h2o, ytterbium sulfate,, ytterbium sulfate octahydrate, ytterbium sulfate, ytterbium iii sulfate octahydrate, reacton, diytterbium 3+ octahydrate trisulfate, sulfuric acid,ytterbium 3+ salt 3:2 , octahydrate 8ci,9ci | |
| MDOSIOHMOWGVIM-UHFFFAOYSA-H | |
| ytterbium(3+);trisulfate;octahydrate | |
| 25022290 | |
| 99.9% | |
| Ytterbium(III) sulfate octahydrate |
| Crystalline | |
| H16O20S3Yb2 | |
| 14,10105 | |
| Soluble in water,moderately soluble in strong mineral acids. | |
| O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 778.40 | |
| 778.38 (634.26 Anhydrous) | |
| Reacton | |
| (REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-727-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only