missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) isopropoxide

Catalog No. p-7050846
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3957903 1 g
AA3957906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3957903 Supplier Thermo Scientific Chemicals Supplier No. 03957903
Only null left

CAS: 6742-69-4 | C9H24O3Yb | 353.342 g/mol

Yttrium(III) tris(isopropoxide) is a catalyst for stereoselective conjugate addition reactions, ring-opening polymerization, generation of reactive enolates, allylic substitution and cyano-phosphorylation of enones and synthesis of nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

CAS 6742-69-4
Molecular Formula C9H24O3Yb
Molecular Weight (g/mol) 353.342
MDL Number MFCD00145532
InChI Key IVGMSSQGEVYWID-UHFFFAOYSA-N
Synonym propan-2-ol; ytterbium, tris isopropanolato ytterbium
PubChem CID 50912003
IUPAC Name propan-2-ol;ytterbium
SMILES CC(C)O.CC(C)O.CC(C)O.[Yb]

Specifications

Melting Point 200°C (decomposition)
Boiling Point 190°C to 200°C (0.2 mmHg)(sublimation)
Odor Odorless
Packaging Ampule
Linear Formula Yb[OCH(CH3)2]3
Quantity 1 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water. Soluble in hexane,toluene,and THF.
Formula Weight 350.31
Physical Form Powder
Chemical Name or Material Ytterbium(III) isopropoxide
Show More Show Less
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA No
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only