missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) Isopropoxide 98.0+%, TCI America™

Catalog No. p-7137985
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y0010200MG 200 mg
Y00101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y0010200MG Supplier TCI America Supplier No. Y0010200MG
Only null left

Chemical Identifiers

CAS 6742-69-4
Molecular Formula C9H24O3Yb
Molecular Weight (g/mol) 353.342
MDL Number MFCD00145532
InChI Key IVGMSSQGEVYWID-UHFFFAOYSA-N
Synonym propan-2-ol; ytterbium, tris isopropanolato ytterbium
PubChem CID 50912003
IUPAC Name propan-2-ol;ytterbium
SMILES CC(C)O.CC(C)O.CC(C)O.[Yb]

Specifications

Melting Point 349°C
Color White-Yellow
Quantity 200 mg
UN Number 2925
Formula Weight 350.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ytterbium(III) Isopropoxide