missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) fluoride, anhydrous, 99.9% (REO)

Catalog No. p-7051521
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4142309 10 g
AA4142318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4142309 Supplier Thermo Scientific Chemicals Supplier No. 04142309
Only null left

For vacuum deposition | CAS: 13760-80-0 | F3Yb | 230.04 g/mol

Ytterbium(III) fluoride is used in laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. It is also used in used in the preparation and study of fluorinated glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13760-80-0
Molecular Formula F3Yb
Molecular Weight (g/mol) 230.04
MDL Number MFCD00049615
InChI Key XASAPYQVQBKMIN-UHFFFAOYSA-K
Synonym ytterbium 3+ trifluoride, ytterbium 3+ ion trifluoride, acmc-209ceg, ytterbium iii ; fluoride, fluoride, fluoride, yb, fluoride, ytterbium iii fluoride, anhydrous trace rare earth metals basis 10g
PubChem CID 21694467
SMILES [F-].[F-].[F-].[Yb+3]

Specifications

Melting Point 1157°C
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids. Insoluble in water.
IUPAC Name ytterbium(3+) trifluoride
Formula Weight 230.04
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 8.2 g/mL
Assay (REO)
Chemical Name or Material Ytterbium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-354-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only