accessibility menu, dialog, popup

UserName

CAS RN 13760-80-0

IUPAC Name: ytterbium(3+) trifluoride
Synonyms: ytterbium(III) fluoride
Molecular Weight (g/mol): 230.04
Molecular Formula: F3Yb
InChi Key: XASAPYQVQBKMIN-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Yb+3]
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)