missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) 2,4-pentanedionate, REacton™, 99.9% (REO)

CAS: 14284-98-1 | C15H21O6Yb | 470.37 g/mol

$143.15 - $515.47

Chemical Identifiers

CAS 14284-98-1
Molecular Formula C15H21O6Yb
Molecular Weight (g/mol) 470.37
MDL Number MFCD00013508
InChI Key JKJCKIFLFXFUKJ-UHFFFAOYSA-N
Synonym ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
PubChem CID 14455610
IUPAC Name (Z)-4-oxopent-2-en-2-olate;ytterbium(3+)
SMILES [Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1315904
View Documents
Thermo Scientific Chemicals
01315904
2 g
Each for $143.15
Only null left
 
AA1315909
View Documents
Thermo Scientific Chemicals
01315909
10 g
Each for $515.47
Only null left
 
Description

Description

Ytterbium(III) 2,4-pentanedionate is used as a catalyst for the preparation of aliphatic oligocarbonate polyols. It takes part in transesterification reactions. It is used for the preparation of polyesters or polyacrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14284-98-1
470.37
JKJCKIFLFXFUKJ-UHFFFAOYSA-N
14455610
[Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
C15H21O6Yb
MFCD00013508
ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
(Z)-4-oxopent-2-en-2-olate;ytterbium(3+)

Specifications

14284-98-1
Odorless
MFCD00013508
ytterbium-2,4-pentanedionate, ytterbium acetyl acetonate
JKJCKIFLFXFUKJ-UHFFFAOYSA-N
(Z)-4-oxopent-2-en-2-olate;ytterbium(3+)
14455610
99.9%
Powder
(REO)
White
C15H21O6Yb
2 g
Insoluble in water.
[Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
470.37
470.37
Reacton
Ytterbium(III) 2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 238-198-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only