accessibility menu, dialog, popup

UserName

YbOOOOOOCCCH3CH3CCH3H3CCH3H3CHHH

CAS RN 14284-98-1

IUPAC Name: ytterbium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: ytterbium(3+) triacetylacetonate
Molecular Weight (g/mol): 470.37
Molecular Formula: C15H21O6Yb
InChi Key: JKJCKIFLFXFUKJ-UHFFFAOYSA-N
SMILES: [Yb+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results