missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human YKL-40 ELISA Kit

Catalog No. BMS2322
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2322 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMS2322 Supplier Invitrogen™ Supplier No. BMS2322

ELISA

The Human YKL-40 (CHI3L1) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human YKL-40.

Human cartilage glycoprotein 39 (GP-39), also known as YKL-40, is a glycoprotein secreted by articular chondrocytes, synoviocytes and macrophages. Serum and synovial fluid GP-39 levels are elevated in inflammatory diseases and correlate with the degree of joint destruction in rheumatoid arthritis. GP-39 is expressed in articular chondrocytes and synovial cells, as well as in liver, but is undetectable in muscle tissues, lung, pancreas, mononuclear cells and fibroblasts. GP-39 is a candidate autoantigen in rheumatoid arthritis and is important in the capacity of cells to respond to and cope with changes in their environment.

Specifications

Accession Number 39 kDa synovial protein; ASRT7; AW208766; Breast regression protein 39; Brp39; BRP39 protein; Cartilage glycoprotein 39; CGP39; CGP-39; Chi3l1; Chil1; chitinase 3 like 1; Chitinase 3 like protein 1 CGP 39; chitinase 3-like 1; chitinase 3-like 1 (cartilage glycoprotein-39); chitinase 3-like 1 protein; chitinase-3-like protein 1; chitinase-like 1; DKFZp686N19119; FLJ38139; Gp39; GP-39; hCGP39; HC-gp39; hCGP-39; HCGP-3 P; YKL40; YKL-40; YYL-40
Assay Range 78 to 5000 pg/mL
Assay Sensitivity 10.83 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1116
Gene Symbol CHI3L1
Interassay CV 7.2%
Intraassay CV 2.3
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 5 μL, Plasma 5 μL, Serum 5 μL
Storage Requirements 2°C to 8°C
Target CHI3L1
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less