missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human YES1, GST Tag Recombinant Protein

Catalog No. A15559
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15559 1 mg
A15557 10 μg
A15558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A15559 Supplier Thermo Scientific™ Supplier No. A15559

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein

For maximum recovery please spin prior to use. Aliquoting the 5 μg, 10 μg, and 20 μg sizes of kinase is not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Active-this protein can be used in kinase activity assays, such as Z'-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA). Purified-purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA). GST-tagged-provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody. Protein Form: Full Length.

YES1 is a recombinant human protein kinase that is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.

Specifications

Accession Number P07947
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 0.02% Triton X-100, 150mM NaCl, 2mM DTT, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 7525
Molecular Weight (g/mol) 88.3 kDa
Name Human YES1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias cellular yes-1 protein; c-Src; c-Yes; EC 2.7.10.2; HsT441; p60c-yes; p60-Src; P61-YES; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; tyrosine-protein kinase Yes; viral oncogene yes homolog; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral (v-yes) oncogene homolog 1; Yamaguchi sarcoma viral oncogene homolog 1; Yes; YES proto-oncogene 1, Src family tyrosine kinase; Yes1; YES1 proto-oncogene, Src family tyrosine kinase
Common Name YES1
Gene Symbol Yes1
Kinase Family SRC Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Tyrosine Kinases
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Full Length
Form Liquid
Research Category Drug Development
Show More Show Less