missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Yellow Elastic Cartilage

Catalog No. S30194
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30194 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30194 Supplier Eisco™ Supplier No. BS18083

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared microscope slide of yellow elastic cartilage. Yellow cartilage is present in outer ear, Eustachian tube and epiglottis. It contains elastic fiber networks and type II collagen fibers.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows charcteristic structures

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Mounting Type Mounting Medium
Type Microbiology slide
For Use With (Application) Microscopy
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Quantity 1
Species Mammal
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less