missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast extract

Catalog No. AC451120250
Change view
Click to view available options
Quantity:
1 kg
5kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451120250 25 kg
AC451120010 1 kg
AC451120050 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC451120250 Supplier Thermo Scientific Chemicals Supplier No. 451120250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84604-16-

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84604-16-

Specifications

Format Fiber Drum
Quantity 25 kg
Chemical Name or Material Yeast Extract
EINECSNumber 283-294-5

RUO – Research Use Only