missing translation for 'onlineSavingsMsg'
Learn More

Xylylazo Violet II 95.0+%, TCI America™

Catalog No. A50061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50061G 1 g
A50065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50061G Supplier TCI America Supplier No. A50061G

Chemical Identifiers

CAS 523-67-1
Molecular Formula C25H21N3O3
Molecular Weight (g/mol) 411.46
MDL Number MFCD00014309
InChI Key ZVNUDNGOWJBORX-NFFVHWSESA-N
Synonym 1-Azo-2-hydroxy-3-(2,4-dimethylcarboxyanilido)naphthalene-1-(2-hydroxybenzene), 2-[2-Hydroxy-3-(2,4-xylylcarbamoyl)-1-naphthylazo]phenol, Xylidyl Blue II
PubChem CID 6832201
IUPAC Name (4Z)-N-(2,4-dimethylphenyl)-4-[2-(2-hydroxyphenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxamide
SMILES CC1=CC=C(NC(=O)C2=CC3=CC=CC=C3\C(=N\NC3=CC=CC=C3O)C2=O)C(C)=C1

Specifications

Melting Point 247°C
Quantity 1 g
Formula Weight 411.46
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Xylylazo Violet II [Spectrophotometric reagent for Mg]