accessibility menu, dialog, popup

UserName

CAS RN 523-67-1

IUPAC Name: (4Z)-N-(2,4-dimethylphenyl)-4-[2-(2-hydroxyphenyl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-2-carboxamide
Synonyms: (4Z)-N-(2,4-dimethylphenyl)-4-[2-(2-hydroxyphenyl)hydrazin-1-ylidene]-3-oxonaphthalene-2-carboxamide
Molecular Weight (g/mol): 411.46
Molecular Formula: C25H21N3O3
InChi Key: ZVNUDNGOWJBORX-NFFVHWSESA-N
SMILES: CC1=CC=C(NC(=O)C2=CC3=CC=CC=C3\C(=N\NC3=CC=CC=C3O)C2=O)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results