missing translation for 'onlineSavingsMsg'
Learn More

Xylose, CP, Spectrum™ Chemical

Catalog No. 18614703
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614703 500 g Poly Pail
18614700 25 g Poly Pail
18614699 100 g Poly Pail
18614698 1 kg Poly Pail
18614702 5 kg Poly Pail
18614701 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614703 Supplier Spectrum Chemical Mfg Cor Supplier No. XY112500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58-86-6 | 150.13 g/mol

Chemical Identifiers

CAS 58-86-6

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 500 g
Molecular Weight (g/mol) 150.13
Grade Pure
Physical Form Powder or Waxy Solid
Chemical Name or Material Xylose

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.