missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol 98.0+%, TCI America™
$32.28 - $136.70
Chemical Identifiers
| CAS | 87-99-0 |
|---|---|
| Molecular Formula | C5H12O5 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00064291,MFCD00064292 |
| InChI Key | HEBKCHPVOIAQTA-UHFFFAOYNA-N |
| Synonym | xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit |
| PubChem CID | 6912 |
| IUPAC Name | pentane-1,2,3,4,5-pentol |
| SMILES | OCC(O)C(O)C(O)CO |
Chemical Identifiers
| 87-99-0 | |
| 152.15 | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| 6912 | |
| OCC(O)C(O)C(O)CO |
| C5H12O5 | |
| MFCD00064291,MFCD00064292 | |
| xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit | |
| pentane-1,2,3,4,5-pentol |
Specifications
| 87-99-0 | |
| White | |
| MFCD00064291,MFCD00064292 | |
| xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit | |
| OCC(O)C(O)C(O)CO | |
| 152.15 | |
| 152.15 | |
| Crystalline Powder |
| 92°C | |
| C5H12O5 | |
| 25 g | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| pentane-1,2,3,4,5-pentol | |
| 6912 | |
| ≥98.0% (T) | |
| Xylitol |
Safety and Handling
EINECSNumber : (9)-0315
RTECSNumber : ZF0800000
TSCA : Yes